missing translation for 'onlineSavingsMsg'
Learn More

Palmitoleic acid, MedChemExpress

Product Code. 16429356
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16429356 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16429356 Supplier Medchem Express Supplier No. HYW011873

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Palmitoleic acid, a composition of fatty acid, is implicated in the prevention of death from cerebrovascular disorders in SHRSP rats.

Palmitoleic acid, a composition of fatty acid, is implicated in the prevention of death from cerebrovascular disorders in SHRSP rats.

Specifications

Chemical Name or Material Palmitoleic acid
CAS 373-49-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C16H30O2
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (393.07 mM; Need ultrasonic)
SMILES CCCCCC/C=C\CCCCCCCC(O)=O
Molecular Weight (g/mol) 254.41
Formula Weight 254.41
Percent Purity 98.0%
Grade Research
Physical Form Liquid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.