missing translation for 'onlineSavingsMsg'
Learn More

Palmitodiolein, MedChemExpress

Product Code. 16470877
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
missing translation for 'unitSize'
10mg
50mg
This item is not returnable. View return policy

Product Code. 16470877

missing translation for 'mfr': Medchem Express HY112132

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Palmitodiolein (Triglyceride POO) is a triacylglycerol which is present in vegetable oils.

Palmitodiolein (Triglyceride POO) is a triacylglycerol which is present in vegetable oils.

Specifications

Chemical Name or Material Palmitodiolein
Physical Form Liquid
CAS 2190-30-9
Color Colorless
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C55H102O6
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym Triglyceride POO Glycerol dioleate palmitate 1,2-Dioleoyl-3-palmitoylglycerol
Solubility Information DMSO : 100 mg/mL (116.36 mM; Need ultrasonic)
SMILES CCCCCCCCCCCCCCCC(OCC(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O)=O
Molecular Weight (g/mol) 859.39
Formula Weight 859.39
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.