missing translation for 'onlineSavingsMsg'
Learn More

Palmitic Acid, TRC

Product Code. 30011587
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011587 10 g 10g
30006942 25 g 25g
30011006 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011587 Supplier Toronto Research Chemicals Supplier No. TRCP14450010G
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-10-3
Molecular Formula C16 H32 O2
Molecular Weight (g/mol) 256.42
Synonym Palmitic acid, Hexadecanoic acid, Palmitic acid (7CI,8CI), 1-Pentadecanecarboxylic acid, A 1600, Cetylic acid, Edenor C 16-98-100, Edenor C16, Emersol 143, FA 1695, Hydrofol Acid 1690, Hystrene 9016, Imex C 1498, Kortacid 1695, Kortacid 1698, Loxiol EP 278, Lunac P 95, Lunac P 95KC, Lunac P 98, NAA 160, NSC 5030, Neo-Fat 16, PA 900, Palmac 98-16, Palmitinic acid, Pentadecanecarboxylic acid, Prifac 2960, Pristerene 4934, V 1695, n-Hexadecanoic acid, n-Hexadecoic acid
IUPAC Name hexadecanoic acid
SMILES CCCCCCCCCCCCCCCC(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)
Formula Weight 256.2402
Percent Purity >95%
Chemical Name or Material Palmitic Acid
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.