missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palmitic Acid-d3, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Palmitic Acid-d3 |
| CAS | 75736-53-7 |
| Molecular Formula | C16 D3 H29 O2 |
| Quantity | 100 mg |
| Synonym | Hexadecanoic-16,16,16-d3 acid (9CI, ACI), [16,16,16-2H3]Hexadecanoic acid, Hexadecanoic-16,16,16-d3 acid, Hexadecanoic acid-16,16,16-d3, Palmitic-16,16,16-d3 acid, Palmitic-ω-d3 acid |
| InChI Formula | InChI=1S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i1D3 |
| SMILES | [2H]C([2H])([2H])CCCCCCCCCCCCCCC(=O)O |
| IUPAC Name | 16,16,16-trideuteriohexadecanoic acid |
| Molecular Weight (g/mol) | 259.44 |
| Formula Weight | 259.2591 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction