missing translation for 'onlineSavingsMsg'
Learn More

Palmitic acid, 95.2%, MP Biomedicals™

Product Code. 11448170 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448170 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11448170 Supplier MP Biomedicals Supplier No. 0210255380
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 57-10-3
Color White
Density 0.853g/mL
Molecular Formula C16H32O2
MDL Number MFCD00002747
Quantity 100 g
Synonym palmitic acid, cetylic acid, palmitate, n-hexadecanoic acid, hexadecylic acid, hydrofol, n-hexadecoic acid, 1-pentadecanecarboxylic acid, palmitinic acid, pentadecanecarboxylic acid
InChI Key IPCSVZSSVZVIGE-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCC(O)=O
IUPAC Name hexadecanoic acid
Molecular Weight (g/mol) 256.43
PubChem CID 985
ChEBI CHEBI:15756
Formula Weight 256.42
Percent Purity 95.2%
Physical Form Crystalline Powder
Chemical Name or Material Palmitic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.