missing translation for 'onlineSavingsMsg'
Learn More

Palmitic Acid, Fisher Chemical

Product Code. 11428702 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11428702 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11428702 Supplier Fisher Chemical Supplier No. P/0280/53

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 57-10-3
Melting Point 59°C
Color White
Boiling Point 351.5°C
Packaging Plastic powder jar
Molecular Formula C16H32O2
MDL Number MFCD00002747
Quantity 500 g
Synonym palmitic acid, cetylic acid, palmitate, n-hexadecanoic acid, hexadecylic acid, hydrofol, n-hexadecoic acid, 1-pentadecanecarboxylic acid, palmitinic acid, pentadecanecarboxylic acid
InChI Key IPCSVZSSVZVIGE-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCC(O)=O
IUPAC Name hexadecanoic acid
Molecular Weight (g/mol) 256.43
PubChem CID 985
ChEBI CHEBI:15756
Formula Weight 256.42g/mol
Physical Form Solid
Show More Show Less
Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.