missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PALMD Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-82018CL1
This item is not returnable.
View return policy
Description
PALMD Polyclonal antibody specifically detects PALMD in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| PALMD | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| C1orf11, palmdelphin, PALMLFLJ20271, Paralemnin-like protein | |
| PALMD antibody was raised against a 15 amino acid peptide near the carboxy terminus of human PALMD. The immunogen is located within amino acids 460 - 510 of PALMD. Amino Acid Squence: QPAKPTPLPRKRSEA | |
| 0.1 mL | |
| Apoptosis, Cancer | |
| 54873 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction