missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PALMD Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18470AF488
This item is not returnable.
View return policy
Description
PALMD Polyclonal antibody specifically detects PALMD in Human samples. It is validated for Western Blot
Specifications
| PALMD | |
| Polyclonal | |
| Western Blot | |
| C1orf11, palmdelphin, PALMLFLJ20271, Paralemnin-like protein | |
| Recognizes region between residues Between between 325 and 375.. | |
| 0.1 mL | |
| Apoptosis, Cancer | |
| 54873 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction