missing translation for 'onlineSavingsMsg'
Learn More

Palmatine Chloride, TRC

Product Code. 30419334
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30419334 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419334 Supplier Toronto Research Chemicals Supplier No. TRCP1442001G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Palmatine chloride
CAS 10605-02-4
Molecular Formula C21 H22 N O4 . Cl
Quantity 1 g
Synonym Dibenzo[a,g]quinolizinium','5,6-dihydro-2,3,9,10-tetramethoxy-','chloride (1:1), Berbinium','7,8,13,13a-tetradehydro-2,3,9,10-tetramethoxy-','chloride (8CI), Dibenzo[a,g]quinolizinium','5,6-dihydro-2,3,9,10-tetramethoxy-','chloride (9CI), Palmatine chloride (6CI,7CI), Fibrauretin
InChI Formula InChI=1S/C21H22NO4.ClH/c1-23-18-6-5-13-9-17-15-11-20(25-3)19(24-2)10-14(15)7-8-22(17)12-16(13)21(18)26-4;/h5-6,9-12H,7-8H2,1-4H3;1H/q+1;/p-1
SMILES [Cl-].COc1cc2CC[n+]3cc4c(OC)c(OC)ccc4cc3c2cc1OC
IUPAC Name 2,3,9,10-tetramethoxy-5,6-dihydroisoquinolino[3,2-a]isoquinolin-7-ium;chloride
Molecular Weight (g/mol) 387.86
Formula Weight 387.1237
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.