missing translation for 'onlineSavingsMsg'
Learn More

Palmatine chloride, MedChemExpress

Product Code. p-200098718
Encompass_Preferred
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 16459895

Brand: Medchem Express HYN0110

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Palmatine chloride is an orally active and irreversible indoleamine 2,3-dioxygenase 1 (IDO-1) inhibitor with IC50s of 3 μM and 157μM against HEK 293-hIDO-1 and rhIDO-1, respectively.

Palmatine chloride can also inhibit West Nile virus (WNV) NS2B-NS3 protease in an uncompetitive manner with an IC50 of 96 μM. Palmatine chloride shows anti-cancer, anti-oxidation, anti-inflammatory, neuroprotection, antibacterial, anti-viral activities.

Specifications

Chemical Name or Material Palmatine chloride
CAS 10605-02-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H22ClNO4
Quantity 250 mg
Solubility Information DMSO : ≥ 32 mg/mL (82.50 mM) ∣H2O : 5 mg/mL (12.89 mM; Need ultrasonic)
SMILES COC1=C(OC)C2=C[N+]3=C(C4=CC(OC)=C(OC)C=C4CC3)C=C2C=C1.[Cl-]
Molecular Weight (g/mol) 387.86
Formula Weight 387.86
Percent Purity 96.21%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.