missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pallidin Antibody (OTI1H9), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73233PECY55
This item is not returnable.
View return policy
Description
Pallidin Monoclonal specifically detects Pallidin in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| Pallidin | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| BLOC1S6 | |
| Full length recombinant protein of human Pallidin (NP_036520) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| OTI1H9 | |
| Flow Cytometry | |
| PALLID, pallid (mouse) homolog, pallidin, Pallid protein homolog, pallidin, pallidin homolog (mouse), PAsyntaxin 13 binding protein 1, Syntaxin 13-interacting protein, syntaxin 13-interacting protein pallid | |
| Mouse | |
| Affinity Purified | |
| Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 26258.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction