missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Key Industrial Equipment™ Pallet box, Geobox, 543L capacity, ventilated side
Brand: Key Industrial Equipment™ A033129
This item is not returnable.
View return policy
Description
High density, injection moulded containers, lightweight and stackable. Plastic pallet boxes are strong, durable, water-resistant and impervious to most chemicals. Stackable, ideal for bulk storage. Ventilation promote airflow. When stacked 10 high the load capacity per container is: 450kg. Easy to clean.- Capacity: 543L
- Internal dimension (H x W x D): 750mm x 1200mm x 910mm
- Two runners on underside
- Weight: 34kg
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction