Learn More
Thermo Scientific Alfa Aesar Palladium wire, 2.0mm (0.08 in.) dia., 99.95% (metals basis)
Brand: Thermo Scientific Alfa Aesar 014761.BQ
Palladium wire, 2.0mm (0.08 in.) dia., 99.95% (metals basis)
Code : NEW
Additional Details : CAS Number : 7440-05-3 Weight : 0.01000kg