missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium Standard: Pd @ 1000 μg/mL in 5% HNO3, VHG™
Click to view available options
Quantity:
50 mL
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
50mL
Description
Palladium Standard: Pd @ 1000 μg/mL in 5% HNO3
Specifications
Specifications
| Chemical Name or Material | Palladium |
| CAS | 5-3-7440 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Palladium |
| Shipping Condition | Room Temperature |
| Molecular Formula | Pd |
| Concentration | 1000 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Palladium on carbon, Palladium Black, Paladio, Palladium/carbon, Palladium, IG 0218A, MPP 030, MPP 050, MPP 080, P 50, P 50 (metal), Palladex 600, Palladium (nanoparticles), Palladium (powder), Palladium element, SFP 1001P, TCP 211 |
| InChI Formula | InChI=1S/Pd |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction