missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium Standard for ICP/MS - Claritas PPT Grade, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
Specifications
Specifications
| CAS | 7440-5-3, 7647-01-0, 7732-18-5 |
| Analyte or Component Names | Palladium Standard |
| Physical Form | Fluid |
| Element | Pd |
| Solvent or Matrix | 2% HCl |
| Quantity | 125 mL |
| Grade | Claritas PPT™ |
| Concentration or Composition (by Analyte or Components) | 1mg/L |
| Chemical Name or Material | Palladium Standard for ICP/MS - Claritas PPT Grade, ICP/MS |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction