missing translation for 'onlineSavingsMsg'
Learn More

Palladium Silver foil, 0.5mm (0.02in) thick, 99.9% (metals basis excluding Pt)

Product Code. 11346139
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Unit Size:
25mm
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11346139 50 x 50 mm Each
11325427 25 x 25 mm 25mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11346139 Supplier Thermo Scientific Alfa Aesar Supplier No. 042684.FI
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Palladium silver foils are used in dentistry for dental crowns and bridges. It also used in conductive films and pastes. It is used to make multilayer capacitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Palladium Silver foil
Name Note 0.5mm (0.02 in.) thick
Avg. Mol. Wt. or Mol. Wt. Range Pd:Ag; 75:25 wt%
Quantity 50 x 50 mm
Odor Odorless
Weight ≈3.64g/25x25mm
Form Foil
Assay Percent Notes (metals basis excluding Pt)
MDL Number MFCD02091738
Solubility Information Insoluble in water.
Percent Purity 99.9%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.