missing translation for 'onlineSavingsMsg'
Learn More

Palladium Silver foil, 0.025mm (0.001in) thick, 99.9% (metals basis excluding Pt)

Product Code. 11356059
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Unit Size:
25mm
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11356059 50 x 50 mm Each
11379837 25 x 25 mm 25mm
11369837 100 x 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11356059 Supplier Thermo Scientific Alfa Aesar Supplier No. 042187.FI
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pd:Ag; 75:25 wt%

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula Pd:Ag; 75:25 wt%
MDL Number MFCD02091738

Specifications

Packaging Tube
Quantity 50 x 50 mm
Percent Purity 99.9%
Chemical Name or Material Palladium Silver foil

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.