missing translation for 'onlineSavingsMsg'
Learn More

Palladium powder, -200 mesh, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog Number 11346418
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
7440-05-3
Pd
106.42
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
7440-05-3
Pd
106.42
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray to Black
Appearance Dark gray to black powder
Assay Percent Range 99.95% (Metals basis)
Quantity 1 g
Mesh Size -200 Mesh
Odor Odorless
Form Powder
Chemical Name or Material Palladium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'