missing translation for 'onlineSavingsMsg'
Learn More

Palladium nitrate & Magnesium nitrate, Matrix Modifier Solution, Specpure™

Product Code. 15463965
Change view
Click to view available options
Quantity:
250 mL
Packaging:
Plastic bottle
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15463965 250 mL Plastic bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15463965 Supplier Thermo Scientific Alfa Aesar Supplier No. 045291.AK
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

0.1% Pd and 0.06% Mg(NO3)2 in 2% HNO3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Plastic bottle
Molecular Formula 0.1% Pd and 0.06% Mg(NO3)2 in 2% HNO3
MDL Number MFCD09910543
Quantity 250 mL
UN Number UN3264
Physical Form Liquid
Chemical Name or Material Palladium nitrate & Magnesium nitrate
compliance-icons
Product Identifier
  • Palladium nitrate & Magnesium nitrate, Matrix Modifier Solution
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: nitric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.