Learn More
Palladium Nickel gauze, 50 mesh woven from 0.1578mm (0.0062in) dia wire, Thermo Scientific Chemicals
Filtration
Brand: Thermo Scientific Alfa Aesar 044698.FL
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Palladium Nickel gauze is used in filtrations. It is also used as a capture gauze for platinum recovery in ammonia oxidation apparatus with a pressure of 4 atm (405 kPa) and a temperature of 8500C.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Palladium Nickel gauze | |
Pd:Ni; 95:5 wt% | |
75 x 75 mm | |
50 Mesh |
50 mesh woven from 0.1578mm (0.0062 in.) dia. wire | |
MFCD05863544 | |
Insoluble in water. | |
Wire Cloth |
RUO – Research Use Only