missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Palladium Molybdenum wire, 0.5mm (0.0197 in.) dia.

Brand:  Thermo Scientific Alfa Aesar 044245.BW

Palladium Molybdenum wire, 0.5mm (0.0197 in.) dia.

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11329778

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers