missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) trifluoroacetate, 97%, Thermo Scientific Chemicals

Catalog Number 11358238
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
42196-31-6
C4F6O4Pd
332.45
MFCD00013204
PBDBXAQKXCXZCJ-UHFFFAOYSA-L
palladium ii trifluoroacetate, palladium ii 2,2,2-trifluoroacetate, palladium 2+ ion ditrifluoroacetate, palladium trifluoroacetate, palladium 2+ ditrifluoroacetate, trifluoroacetic acid palladium ii salt, bis 2,2,2-trifluoroacetoxy palladium, acmc-209jo7, ksc235m3d
2733347
[Pd++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F
This item is not returnable. View return policy
42196-31-6
C4F6O4Pd
332.45
MFCD00013204
PBDBXAQKXCXZCJ-UHFFFAOYSA-L
palladium ii trifluoroacetate, palladium ii 2,2,2-trifluoroacetate, palladium 2+ ion ditrifluoroacetate, palladium trifluoroacetate, palladium 2+ ditrifluoroacetate, trifluoroacetic acid palladium ii salt, bis 2,2,2-trifluoroacetoxy palladium, acmc-209jo7, ksc235m3d
2733347
[Pd++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F

Palladium(II) trifluoroacetate is used in the preparation of allyl-palladium complexes. It acts as a catalyst used in the mild decarboxylation of electron-rich aromatic acids and the direct cross-coupling of unactivated arenes. Further, it is involved in the selective allylic oxidation of geranylacetone and other olefins to their allyl acetates in order to prepare keto alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 210°C (decomposition)
Color Brown
Physical Form Powder
Quantity 1 g
Assay Percent Range 97%
Solubility Information Soluble in diethyl ether and acetone. Insoluble in benzene,chloroform and trifluoroacetic acid.
IUPAC Name palladium(2+) ditrifluoroacetate
Formula Weight 332.44
Sensitivity Hygroscopic
Appearance Brown powder
Chemical Name or Material Palladium(II) trifluoroacetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'