missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium(II) trifluoroacetate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Palladium(II) trifluoroacetate is used in the preparation of allyl-palladium complexes. It acts as a catalyst used in the mild decarboxylation of electron-rich aromatic acids and the direct cross-coupling of unactivated arenes. Further, it is involved in the selective allylic oxidation of geranylacetone and other olefins to their allyl acetates in order to prepare keto alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 210°C (decomposition) |
Color | Brown |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 97% |
Solubility Information | Soluble in diethyl ether and acetone. Insoluble in benzene,chloroform and trifluoroacetic acid. |
IUPAC Name | palladium(2+);2,2,2-trifluoroacetate |
Formula Weight | 332.44 |
Sensitivity | Hygroscopic |
Appearance | Brown powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'