missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) sulfate, solution, Pd 6% w/w (cont. Pd), Thermo Scientific™

Product Code. 11490290
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490290 5 g 5g
11400300 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490290 Supplier Thermo Scientific Alfa Aesar Supplier No. 012622.NB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13566-03-5
Molecular Formula O4PdS
Molecular Weight (g/mol) 202.48
MDL Number MFCD00011173
InChI Key RFLFDJSIZCCYIP-UHFFFAOYSA-L
Synonym palladium ii sulfate, palladium sulfate, palladium 2+ sulfate, palladium sulphate, palladous sulfate, palladium monosulfate, palladium sulfate pdso4, sulfuric acid, palladium salt, acmc-20ajzn, pdso4
PubChem CID 166846
IUPAC Name palladium(2+);sulfate
SMILES [Pd++].[O-]S([O-])(=O)=O

Specifications

Color Red to Brown
Linear Formula PdSO4
UN Number UN3264
Concentration Pd 6% w/w (cont. Pd)
Solubility Information Fully miscible with water.
Formula Weight 202.48 (Anhydrous)
Quantity 5 g
Physical Form Liquid
Chemical Name or Material Palladium(II) sulfate, Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.