missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) oxide monohydrate, 99.9% (metals basis), Pd 73% min

Product Code. p-7048621
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11341678

Brand: Thermo Scientific Alfa Aesar 011041.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 64109-12-2 | H2O2Pd | 140.43 g/mol

Palladium(II) oxide monohydrate is used as an Hydrogenation catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64109-12-2
Molecular Formula H2O2Pd
Molecular Weight (g/mol) 140.43
MDL Number MFCD00011172
InChI Key CLKURSCFRVRBKZ-UHFFFAOYSA-N
Synonym palladium oxide hydrate, opd.h2o, palladium oxide, hydrate, oxopalladium hydrate, oxopalladium-water 1/1, palladium ii oxide hydrate, palladium ii oxide monohydrate, palladium ii oxide hydrate pd basis, palladium ii oxide hydrate, pd content ∼97 %, palladium ii oxide hydrate trace metals basis
PubChem CID 16211505
SMILES O.[O--].[Pd++]

Specifications

Melting Point (decomposition)
Physical Form Powder
Quantity 2 g
Merck Index 14,6993
Solubility Information Insoluble in water.
IUPAC Name oxopalladium;hydrate
Formula Weight 140.41 (122.40 Anhydrous)
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Pd 73% min.
Assay (metals basis)
Chemical Name or Material Palladium(II) oxide monohydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.