missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-2-propanol, 98+%

Product Code. 11413067
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413067 500 g 500g
11493057 25 g 25g
11403067 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413067 Supplier Thermo Scientific Alfa Aesar Supplier No. A15580.36
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 617-94-7 | C9H12O | 136.19 g/mol

It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as chemical intermediate.

Solubility
Insoluble in water.

Notes
Store away from acids, acid chlorides, acid anhydrides, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-94-7
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00004456
InChI Key BDCFWIDZNLCTMF-UHFFFAOYSA-N
Synonym 2-phenyl-2-propanol, dimethylphenylcarbinol, 1-hydroxycumene, dimethylphenylmethanol, alpha-cumyl alcohol, phenyldimethylcarbinol, 2-propanol, 2-phenyl, 2-phenylisopropanol, alpha,alpha-dimethylbenzyl alcohol, 2-phenyl-propan-2-ol
PubChem CID 12053
IUPAC Name 2-phenylpropan-2-ol
SMILES CC(C)(O)C1=CC=CC=C1

Specifications

Melting Point 29°C to 34°C
Density 0.973
Boiling Point 215°C
Flash Point 87°C (188°F)
Refractive Index 1.5196
Quantity 500 g
Beilstein 1905012
Solubility Information Insoluble in water.
Formula Weight 136.19
Percent Purity ≥98%
Chemical Name or Material 2-Phenyl-2-propanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.