missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) oxide, 99.995%, (trace metal basis)

Product Code. p-4258697 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10804161

Brand: Thermo Scientific Acros 193680010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1314-08-5 | OPd | 122.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-08-5
Molecular Formula OPd
Molecular Weight (g/mol) 122.42
MDL Number MFCD00011172
InChI Key JQPTYAILLJKUCY-UHFFFAOYSA-N
Synonym palladium ii oxide, palladium oxide pdo, acmc-1bqvh, palladium oxide, pdo, palladium ii oxide, anhydrous, palladium ii oxide-pd, 11113-77-2 cpd with unspecified mf, palladium oxide ? inverted exclamation mark-?, palladium ii oxide trace metals basis
PubChem CID 5083724
IUPAC Name palladium(2+) oxidandiide
SMILES [O--].[Pd++]

Specifications

Melting Point 870.0°C
Physical Form Viscous Liquid
Quantity 1 g
Assay Percent Range 85.2 to 88.7% (Pd)
Linear Formula PdO
Merck Index 15,7094
Solubility Information Solubility in water: practically insoluble. Other solubilities: practically insoluble in acids, slightly soluble in aqua regia, soluble in 48% hbr
Formula Weight 122.4
Percent Purity 99.995%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Palladium(II) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.