missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) nitrate, solution, Pd 4-5% w/w (cont. Pd), Thermo Scientific Chemicals

Product Code. 15432327
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15432327 25 g 25g
15426985 1 g 1g
15416985 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15432327 Supplier Thermo Scientific Alfa Aesar Supplier No. 045512.ND
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Palladium(II) nitrate dehydrate is used as a reactant for doping activated carbon for catalysis. It acts as a catalyst for hydrogenation of 1-heptyne and naphthalene. It serves as an important raw material for the preparation of platinum-palladium carbon alloy nanocatalysts for methanol-tolerant oxygen reduction reaction in fuel cells and in the synthesis of copper palladium alloy thin films. Furthermore, it is used in the synthesis of di-phenyl sulfide-modified palladium-titanium dioxide catalysts for acetylene hydrogenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-05-3
Molecular Formula N2O6Pd
Molecular Weight (g/mol) 230.43
MDL Number MFCD00011169
InChI Key GPNDARIEYHPYAY-UHFFFAOYSA-N
Synonym palladium nitrate, palladium ii nitrate, palladous nitrate, palladium dinitrate, unii-5g27lbz05u, nitric acid, palladium 2+ salt, hydrogen tetranitropalladate ii, palladium 2+ dinitrate, palladiumdinitrate
PubChem CID 24932
IUPAC Name palladium(2+);dinitrate
SMILES [Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Brown to Orange
Linear Formula Pd(NO3)2
UN Number UN3264
Concentration Pd 4 to 5% w/w (cont. Pd)
Solubility Information Miscible with water and dilute nitric acid.
Formula Weight 230.43
Quantity 25 g
Physical Form Liquid
Chemical Name or Material Palladium (II) nitrate solution

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.