Learn More
Palladium(II) nitrate, solution, Pd 12-16% w/w (cont. Pd), Thermo Scientific™
£173.00 - £660.00
Chemical Identifiers
CAS | 10102-05-3 |
---|---|
Molecular Formula | N2O6Pd |
Molecular Weight (g/mol) | 230.43 |
MDL Number | MFCD00011169 |
InChI Key | GPNDARIEYHPYAY-UHFFFAOYSA-N |
Synonym | palladium nitrate, palladium ii nitrate, palladous nitrate, palladium dinitrate, unii-5g27lbz05u, nitric acid, palladium 2+ salt, hydrogen tetranitropalladate ii, palladium 2+ dinitrate, palladiumdinitrate |
PubChem CID | 24932 |
IUPAC Name | palladium(2+);dinitrate |
SMILES | [Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
10102-05-3 | |
230.43 | |
GPNDARIEYHPYAY-UHFFFAOYSA-N | |
24932 | |
[Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
N2O6Pd | |
MFCD00011169 | |
palladium nitrate, palladium ii nitrate, palladous nitrate, palladium dinitrate, unii-5g27lbz05u, nitric acid, palladium 2+ salt, hydrogen tetranitropalladate ii, palladium 2+ dinitrate, palladiumdinitrate | |
palladium(2+);dinitrate |
Specifications
Brown to Orange | |
N2O6Pd | |
UN3264 | |
palladium nitrate, palladium ii nitrate, palladous nitrate, palladium dinitrate, unii-5g27lbz05u, nitric acid, palladium 2+ salt, hydrogen tetranitropalladate ii, palladium 2+ dinitrate, palladiumdinitrate | |
GPNDARIEYHPYAY-UHFFFAOYSA-N | |
palladium(2+);dinitrate | |
24932 | |
Liquid |
10102-05-3 | |
MFCD00011169 | |
Pd 12 to 16% w/w (cont. Pd) | |
Soluble in water,nitric acid. | |
[Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
230.43 | |
230.43 | |
Palladium(II) nitrate, Solution |
Safety and Handling
GHS H Statement
H314-H318-H290-H302-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May be corrosive to metals.
Harmful if swallowed.
May cause an allergic skin reaction.
P234-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
H290-H302+H332-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 233-265-8
missing translation for 'rtecsNumber' : QV0560000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures