missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) chloride, solution, Pd 20-25% w/w (cont. Pd), Thermo Scientific™

Catalog Number 11460241
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
7647-10-1
Cl2Pd
177.32
MFCD00003558
PIBWKRNGBLPSSY-UHFFFAOYSA-L
palladium chloride, palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium chloride pdcl2, enplate activator 440
24290
CHEBI:53434
dichloropalladium
[Cl-].[Cl-].[Pd++]
This item is not returnable. View return policy
7647-10-1
Cl2Pd
177.32
MFCD00003558
PIBWKRNGBLPSSY-UHFFFAOYSA-L
palladium chloride, palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium chloride pdcl2, enplate activator 440
24290
CHEBI:53434
dichloropalladium
[Cl-].[Cl-].[Pd++]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Brown
UN Number UN3264
Concentration Pd 20 to 25% w/w (cont. Pd)
Solubility Information Soluble in aqueous metal halides,water,acetone,alcohol,hydrobromic acid and hydrochloric acid.
Formula Weight 177.31
Quantity 5 g
Physical Form Liquid
Chemical Name or Material Palladium(II) chloride Solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'