missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) chloride, solution, Pd 20-25% w/w (cont. Pd), Thermo Scientific™

Product Code. p-7050956
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460241 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11460241 Supplier Thermo Scientific Alfa Aesar Supplier No. 040019.NB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-10-1
Molecular Formula Cl2Pd
Molecular Weight (g/mol) 177.32
MDL Number MFCD00003558
InChI Key PIBWKRNGBLPSSY-UHFFFAOYSA-L
Synonym palladium chloride, palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium chloride pdcl2, enplate activator 440
PubChem CID 24290
ChEBI CHEBI:53434
IUPAC Name dichloropalladium
SMILES [Cl-].[Cl-].[Pd++]

Specifications

Color Brown
UN Number UN3264
Concentration Pd 20 to 25% w/w (cont. Pd)
Solubility Information Soluble in aqueous metal halides,water,acetone,alcohol,hydrobromic acid and hydrochloric acid.
Formula Weight 177.31
Quantity 5 g
Physical Form Liquid
Chemical Name or Material Palladium(II) chloride Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.