missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) bromide, Premion™, 99.998% (metals basis), Pd 39.5% min

Product Code. 11320288
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11320288 1 g 1g
11310288 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11320288 Supplier Thermo Scientific Alfa Aesar Supplier No. 043697.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13444-94-5 | Br2Pd | 266.23 g/mol

Palladium(II) bromide is used as a source of the palladium(0) precatalyst in various coupling reactions. It is used as a catalyst for the carbonylative Heck coupling reaction. It is also used as an oxidizing agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13444-94-5
Molecular Formula Br2Pd
Molecular Weight (g/mol) 266.23
MDL Number MFCD00011170
InChI Key INIOZDBICVTGEO-UHFFFAOYSA-L
Synonym palladium ii bromide, palladium bromide, palladium bromide pdbr2, palladous bromide, palladium ii bromide, premion, palladium 2+ dibromide, pdbr2, palladiumbromide, palladiumbromide pdbr2
PubChem CID 83469
IUPAC Name dibromopalladium
SMILES [Br-].[Br-].[Pd++]

Specifications

Melting Point (decomposition)
Color Black
Physical Form Powder
Quantity 1 g
Solubility Information Insoluble in water. Soluble in hydrogen bromide.
Formula Weight 266.22
Percent Purity 99.998%
Grade Premion
Concentration or Composition (by Analyte or Components) Pd 39.5% min
Assay (metals basis)
Chemical Name or Material Palladium (II) bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.