missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) Acetate, TRC

Product Code. 30428214
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
Product Code. Quantity unitSize
30428214 50 mg -
30410945 100 mg 100mg
30414360 250 mg -
3 options
This item is not returnable. View return policy

Product Code. 30428214

Brand: Toronto Research Chemicals TRCP14110550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3375-31-3
Formula Weight 223.9301
InChI Formula InChI=1S/2C2H4O2.Pd/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2
IUPAC Name palladium(2+);diacetate
Molecular Formula (C2H3O2)2Pd
Molecular Weight (g/mol) 224.51
SMILES [Pd+2].CC(=O)[O-].CC(=O)[O-]
Synonym Palladium(2+) Acetic Acid Salt, Palladium Acetic Acid Salt, Bis(Acetato)palladium, Diacetatopalladium, Diacetoxypalladium, HyCat 1, HyCat Base 1, Palladium Diacetate, Palladous Acetate
Chemical Name or Material Palladium(II) Acetate

Specifications

Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.