missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) acetate, Pd 45.9-48.4%

Product Code. 11356028
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11356028 5 g 5g
11346028 1 g 1g
11336028 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11356028 Supplier Thermo Scientific Alfa Aesar Supplier No. 010516.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3375-31-3 | C4H6O4Pd | 224.51 g/mol

Palladium(II)acetate is a catalyst of choice for a wide variety of reactions such as vinylation, Wacker process, Buchwald-Hartwig amination, carbonylation, oxidation, rearrangement of dienes (e.g., Cope rearrangement), C-C bond formation, reductive amination, etc. Precursor to Pd(0), other Pd(II) compounds of catalytic significance, and Pd nanowires.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3375-31-3
Molecular Formula C4H6O4Pd
Molecular Weight (g/mol) 224.51
MDL Number MFCD00012453
InChI Key YJVFFLUZDVXJQI-UHFFFAOYSA-L
Synonym palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate
PubChem CID 167845
IUPAC Name palladium(2+);diacetate
SMILES [Pd++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point ∽205°C (decomposition)
Color Red to Brown
Physical Form Needles
Quantity 5 g
Assay Percent Range 45.9 to 48.4%
Solubility Information Soluble as monomer in glacial acetic acid or as trimer in benzene.
Formula Weight 673.46
Appearance Various forms in red-brown color (powder/flake/crystalline/beads)
Chemical Name or Material Palladium(II) acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.