missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium(II) acetate, 99.9%, (trace metal basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
Specifications
Melting Point | 205.0°C |
Quantity | 5 g |
Assay Percent Range | 46.9 to 48.5% (Pd) |
Linear Formula | (CH3CO2)2Pd |
Merck Index | 15,7092 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, methylene dichloride,, chloroform, acetonitrile and diethylether, insoluble in alcohols |
Formula Weight | 224.51 |
Percent Purity | 99.9% |
Grade | Trace Metal Basis |
Packaging | Glass bottle |
Show More |
Safety and Handling
Product Identifier
- Palladium(II) acetate
- Danger
- Serious eye damage/eye irritation Category 1
- LONG-TERM AQUATIC HAZARD Chronic 4
- H318-Causes serious eye damage.
- H413-May cause long lasting harmful effects to aquatic life.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'