missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) 2,4-pentanedionate, Pd 34.7%

Product Code. 11366028
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11366028 5 g 5g
11376028 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11366028 Supplier Thermo Scientific Alfa Aesar Supplier No. 010517.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Carbonylation, oligomerization | CAS: 14024-61-4 | C10H14O4Pd | 304.64 g/mol

It is used in carbonylation, oligomerization, and as a catalyst precursor for various synthetic organic transformations, for example in the preparation of complexes to efficiently catalyze Heck reaction. Useful in the decarboxylative cross-coupling of arylcarboxylic acids with aryl halides, in the preparation of monodisperse nanoparticles of CuPd alloy and in several cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14024-61-4
Molecular Formula C10H14O4Pd
Molecular Weight (g/mol) 304.64
MDL Number MFCD00000025 MFCD00000025
InChI Key JKDRQYIYVJVOPF-FDGPNNRMSA-L
Synonym palladium diacetonate, acetylacetone palladium ii salt, bis 2,4-pentanedionato palladium ii
PubChem CID 53384484
IUPAC Name (E)-4-oxopent-2-en-2-olate;palladium(2+)
SMILES [Pd++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 210°C (decomposition)
Color Yellow to Orange
Physical Form Powder
Quantity 5 g
Solubility Information Soluble in benzene and chloroform.
Formula Weight 304.92
Concentration or Composition (by Analyte or Components) Pd 34.7%
Odor Odorless
Chemical Name or Material Palladium(II) 2,4-pentanedionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.