missing translation for 'onlineSavingsMsg'
Learn More

Palladium hydroxide, 99%

Product Code. 11338298
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338298 0.25 g 0.25g
11348298 1 g 1g
11358298 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11338298 Supplier Thermo Scientific Alfa Aesar Supplier No. 039496.01
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12135-22-7 | H2O2Pd | 140.43 g/mol

Acts as a catalyst for arylation reactions. It is employed in hydrogenolysis of benzyl-nitrogen bonds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12135-22-7
Molecular Formula H2O2Pd
Molecular Weight (g/mol) 140.43
MDL Number MFCD00064599
InChI Key NXJCBFBQEVOTOW-UHFFFAOYSA-L
Synonym palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon
PubChem CID 9942167
SMILES [OH-].[OH-].[Pd++]

Specifications

Color Black
Physical Form Powder
Quantity 0.25 g
Assay Percent Range 99%
Solubility Information Insoluble in water.
IUPAC Name palladium(2+);dihydroxide
Formula Weight 140.42
Percent Purity 99%
Odor Odorless
Chemical Name or Material Palladium hydroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.