missing translation for 'onlineSavingsMsg'
Learn More

Palladium black, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11386607
Click to view available options
Quantity:
1 g
5 g

Use as a catalyst in hydrogenation reactions. Other uses include the manufacture of paints, adhesives, fibers and coatings. Basic particles of Palladium black have the tendency to cluster into macroscopic aggregates.

It is used as a catalyst in hydrogenation reactions. Further it finds use in the manufacture of paints, adhesives, fibers and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-05-3
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]

Specifications

Color Gray to Silver
Melting Point 1555°C
Boiling Point 3067°C
Appearance Powder
Density 12.02g/cm3 at 20°C
Assay Percent Range 99.9% (Metals basis)
Quantity 1 g
Solubility Information Insoluble in water; Soluble in aqua regia and fused alkalies
Odor Odorless
Form Powder
Chemical Name or Material Palladium black
Show More Show Less
compliance-icons
Product Identifier
  • Palladium black
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P370+P378-In case of fire: Use to extinguish.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'