missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium on barium sulfate, 5% Pd, unreduced, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
50 g
Packaging:
Glass bottle
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Color | Brown |
Quantity | 10 g |
Fieser | 01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356 |
Merck Index | 15, 7090 |
Formula Weight | 106.42 |
Percent Purity | 4.50 to 5.50% (Pd) |
Packaging | Glass bottle |
Form | Powder |
Chemical Name or Material | Palladium on barium sulfate, 5% Pd |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'