missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Palladium API Fluorescent Detection Kit

Product Code. 15800913
Click to view available options
Quantity:
96 Tests
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 15800913

Brand: Invitrogen™ EIAPDAPIF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

96-well plates, standard, other assay specific reagents, buffers and lot-specific technical data sheet.

Assay Palladium API Activity in various solvents including toluene, ethanol, acetonitrile, DMSO and DMF containing samples.

The Palladium Detection research-use-only kit is a fluorescent assay designed for the quantification and detection of palladium in various solvents including toluene, ethanol, acetonitrile, DMSO and DMF containing samples.

This complete, ready-to-use kit includes black 96-well plate(s), palladium standard, palladium detection reagent, and other components to perform the assay. A 96-well microplate reader capable of reading fluorescent emission at 520 nm, with excitation at 485 nm, is required for use of this kit.

Performance characteristics
• Assay type: fluorescent detection kit
• Sample types: various solvents including toluene, ethanol, acetonitrile, DMSO, and DMF
• Standard curve range: 3.1–100 nM
• Reactivity: species independent

Background
Many new synthetic transformations that have been developed in recent years use palladium (Pd) compounds for the catalysis of carbon-carbon and carbon-heteroatom coupling reactions such as the Buchwald-Hartwig, Heck, Kumada, Negishi, Nozaki-Hiyama, Sonogashira, Stille, Suzuki-Miyaura, and Tsuji-Trost transformations. These reactions have found increased popularity for pharmaceutical processes as they utilize a wide-range of functional groups and can therefore be used to build complicated molecules. However, palladium-catalyzed reactions present a problem in that the palladium can often be retained in the isolated product.

Assay principle
The standard methods of quantifying palladium in samples are atomic absorption analysis, x-ray fluorescence, and plasma emission spectroscopy such as inductively-coupled plasma mass spectroscopy (ICP-MS). These methods require expensive instruments with a highly trained scientist to operate. Cross-contamination of the instrument can limit the throughput of this analysis and requires scrupulous clean up methodology.

This Palladium Fluorescent Detection Kit is designed to allow the rapid determination of the relative amounts of Pd present in a variety of samples and solvents including toluene, ethanol, acetonitrile, DMSO, and DMF. The kit uses a non-fluorescent detection molecule that, in the presence of palladium, cleaves to yield a brightly fluorescent product.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 96-well plate(s), standard, other assay specific reagents, buffers, and lot-specific technical data sheet.

Store at –20°C.
Immunoassay Kit Format Fluorescent Detection Kit
For Use With (Equipment) Microplate Reader
Gene ID (Entrez) Non-Gene
Gene Symbol Non-Gene
Quantity 96 Tests
Shipping Condition Approved for shipment on Wet or Dry Ice
Target Kit Named Palladium API

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.