missing translation for 'onlineSavingsMsg'
Learn More

Palladium on activated carbon, 10% Pd, unreduced, dry, Thermo Scientific Chemicals

Catalog Number 10073163
Click to view available options
Quantity:
1 g
10 g
50 g
250 g
Packaging:
Glass bottle
7440-05-3, 7440-44-0
Pd
106.42
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
7440-05-3, 7440-44-0
Pd
106.42
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Black
Quantity 1 g
Fieser 01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356
Merck Index 15, 7090
Formula Weight 106.42
Percent Purity 10% (Pd)
Packaging Glass bottle
Particle Size Particle size distribution: ≤4 Micron: 10%, ≤21 Micron: 50%, ≤90 Micron: 90%
Form Powder
Chemical Name or Material Palladium on activated carbon, 10% Pd
compliance-icons
Product Identifier
  • Palladium on activated carbon, unreduced
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P233-Keep container tightly closed.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'