missing translation for 'onlineSavingsMsg'
Learn More

Palladium, AAS standard solution, Specpure™ Pd 1000μg/mL, Thermo Scientific Chemicals

Product Code. 11396359
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396359 500 mL 500mL
11329179 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11396359 Supplier Thermo Scientific Alfa Aesar Supplier No. 088085.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD03613603

Specifications

Physical Form Liquid
Concentration Pd in 20% HCl
UN Number UN1789
Quantity 500 mL
Grade Specpure
Chemical Name or Material Palladium, AAS standard solution, Pd 1000μg/mL

RUO – Research Use Only

Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.