missing translation for 'onlineSavingsMsg'
Learn More

Palladium, 5% on barium sulfate powder, unreduced, Thermo Scientific Chemicals

Catalog Number 11378007
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
7440-05-3
Pd
106.42
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
This item is not returnable. View return policy
7440-05-3
Pd
106.42
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]

Hydrodehalogenation, Rosenmund reduction, asymetric hydrogenation

Hydrodehalogenation, Rosenmund reduction, asymetric hydrogenationPalladium, 5% on barium sulfate powder is used as a catalyst for hydrogenation reaction. It is also used to catalyze the hydrogenation of acyl chlorides to aldehydes. Further, it is used in the Rosenmund reduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown
Quantity 10 g
Solubility Information Insoluble in water.
Chemical Name or Material Palladium, 5% on barium sulfate powder, Unreduced

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'