missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Palladium, 4% Platinum, 1% on carbon paste E101023-4/1

Brand:  Thermo Scientific Alfa Aesar H37009.22

Palladium, 4% Platinum, 1% on carbon paste E101023-4/1

Code : NEW

Additional Details : Weight : 0.10000kg

Product Code. 15409095

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers