missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Palladium, 3% on activated carbon powder, standard, reduced, nominally 50% water wet
Brand: Alfa Aesar™ 038296.06
Code : NEW
Additional Details : Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
Palladium | |
3% on activated carbon powder, Standard, reduced, nominally 50% water wet | |
MFCD03457879 |
Specifications
Palladium | |
MFCD03457879 | |
Odorless |
3% on activated carbon powder, Standard, reduced, nominally 50% water wet | |
5g | |
900 to 1100m2/g |