missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Palladium, 10% on activated carbon paste, Type 39

Brand:  Alfa Aesar™ H37431.22

Code : NEW

Additional Details : CAS Number : 7440-05-3 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 15479095

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Palladium
7440-05-3
10% on activated carbon paste, Type 39
MFCD03457879

Specifications

Palladium
7440-05-3
MFCD03457879
Dark gray
10% on activated carbon paste, Type 39
Solid
Odorless
100g
Videos