missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar Palladium, 1% on 1-2 mm alumina spheres, reduced

Brand:  Alfa Aesar 044820.22

Palladium, 1% on 1-2 mm alumina spheres, reduced

Code : NEW

Additional Details : Weight : 0.10000kg

Product Code. 11440121

  • £286.20 / 100g


Estimated Shipment date
from Supplier 25-04-2018
Add to basket