missing translation for 'onlineSavingsMsg'
Learn More

Palladium, 0.5% on 3.18mm (0.125in) alumina pellets, unreduced, Thermo Scientific Chemicals

Catalog Number 11339389
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
7440-05-3
Pd
106.42
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
This item is not returnable. View return policy
7440-05-3
Pd
106.42
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]

Palladium, 0.5% on 3.18mm (0.125in) alumina pellets is a heterogeneous catalyst used in chemical processes for reactions ranging from gas phase oxidation through selective hydrogenation of chemical, petrochemical, and pharmaceutical feedstocks to fuel cell power generation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 100 g
Solubility Information Insoluble in water.
Chemical Name or Material Palladium, 0.5% on alumina pellets, 3.18mm (0.125 in.), Unreduced

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'