missing translation for 'onlineSavingsMsg'
Learn More

Palladin Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen Palladin
Concentration 0.21 mg/mL
Applications Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Flow Cytometry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16897513
View Documents
Proteintech
10853-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16887513
View Documents
Proteintech
10853-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody, generated against the fragment 999-1383 aa of palladin, recognizes most isoforms of this protein, except isoform 6.

Palladin is a cytoskeletal protein required for organization of normal actin cytoskeleton. Palladin plays roles in establishing cell morphology, motility, cell adhesion and cell-extracellular matrix interactions in a variety of cell types. Palladin may function as a scaffolding molecule with the potential to impact both actin polymerization as well as the assembly of existing actin filaments into higher-order arrays. It binds to proteins that bind to either monomeric or filamentous actin. It localizes at sites where active actin remodeling takes place, such as lamellipodia and membrane ruffles. Palladin′s different isoforms may have functional differences. It is also involved in the control of morphological and cytoskeletal changes associated with dendritic cell maturation.
Specifications

Specifications

Palladin
Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Flow Cytometry
Unconjugated
Rabbit
Human, Mouse
Q8WX93, Q9ET54
23022, 72333
Palladin Fusion Protein Ag1288
Primary
-20°C
PALLD
0.21 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
CGI 151, KIAA0992, palladin, PALLD, PNCA1, Sarcoma antigen NY SAR 77, SIH002
Palld
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.