Learn More
Palladin Rabbit anti-Human, CoraLite 555, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL555-10853-100UL
Description
The Palladin antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human Palladin. This antibody recognizes human, mouse antigen. The Palladin antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™555.
Specifications
| Palladin | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Mouse |
| Flow Cytometry (Intracellular Staining) | |
| CoraLite 555 | |
| 100 μL | |
| 23022 | |
| IgG |